missing translation for 'onlineSavingsMsg'
Learn More

Ansell Nitritex™ Omega Pro-tech™ Disposable Powder-free Examination Gloves

Product Code. 12980835
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Purple nitrile exam gloves resist cuts and snags and are available in a variety of sizes.

  • Purple
  • Beaded cuff
  • Non-sterile
  • Nitrile contains no proteins
  • Various sizes
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Complies with the requirements of the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3), Glove Standard EN 420:2003 & EN 374 (Parts 1, 2 & 3:2003), ISO 9001:2008 & 13485:2003, 93/42/EEC Annex V and 89/686/EEC Article 11b certifications, Class I Medical Device conforming to MDD Directive 93/42/EEC and Category 3 - Complex Design Personal Protective Equipment conforming to PPE Directive 89/686/EEC, CE 0120
Classification PPE category III
Color Purple
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
Sterility Non-sterile
Finger Style Textured
Glove Cuff Style Beaded/Rolled
Inspected for Medical Use Yes
Quantity 100/Pk.
Resistant To Cuts and snags, wide range of chemicals
Series OPT
Length (Metric) 24 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'