Learn More
Ohaus™ OIML F2 Weight Set
Adjusted to Meet or Exceed OIML F2 Tolerances
313.00 € - 1940.00 €
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16622212
|
Ohaus™
30390829 |
500 mg to 1 mg |
313.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16612212
|
Ohaus™
30390827 |
50 g to 1 mg |
564.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16602212
|
Ohaus™
30390826 |
50 g to 1 g |
328.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16692202
|
Ohaus™
30390825 |
100 g to 1 mg |
621.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16682202
|
Ohaus™
30390824 |
100 g to 1 g |
390.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16632192
|
Ohaus™
30390032 |
2 x 200 g to 1 mg |
759.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16622192
|
Ohaus™
30390031 |
2 x 200 g to 1 g |
523.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16672202
|
Ohaus™
30390821 |
500 g to 1 mg |
887.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16662202
|
Ohaus™
30390820 |
500 g to 1 g |
621.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16652202
|
Ohaus™
30390819 |
1 kg to 1 mg |
1360.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16642202
|
Ohaus™
30390818 |
1 kg to 1 g |
1105.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16612192
|
Ohaus™
30390030 |
2 x 2 kg to 1 mg |
1730.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16602192
|
Ohaus™
30390029 |
2 x 2 kg to 1 g |
1530.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16632202
|
Ohaus™
30390815 |
5 kg to 1 mg |
1940.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
16622202
|
Ohaus™
30390814 |
5 kg to 1 g |
1710.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
Description
- Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
Protective Case |