missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ OIML Class F1 Proof Line Knob Weight without Certificate

Polished non-magnetizable calibration weights. Sartorius™ OIML Class F1 Knob Weight without Certificate is constructed of special steel and supplied in a plastic case.

57.65 € - 1180.00 €

Specifications

Product Type Proof Line Weight
Classification OIML Class F1
Material Special Stainless Steel
Density 7,950kg/m3
View More Specs

Products 13
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
11383294
View Documents Promotion Details
Sartorius™
YCW313-AC-00
1 g
57.65 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
11393294
View Documents Promotion Details
Sartorius™
YCW323-AC-00
2 g
58.65 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
11303304
View Documents Promotion Details
Sartorius™
YCW353-AC-00
5 g
61.95 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
11313304
View Documents Promotion Details
Sartorius™
YCW413-AC-00
10 g
68.30 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
11323304
View Documents Promotion Details
Sartorius™
YCW423-AC-00
20 g
74.65 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
11333304
View Documents Promotion Details
Sartorius™
YCW453-AC-00
50 g
95.95 €
Each
Estimated Shipment: 16-12-2024
to see stock.
Añadir a la cesta
 
11343304
Ver documentos Detalles de la promoción
Sartorius™
YCW513-AC-00
100 g
113.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Añadir a la cesta
 
11353304
Ver documentos Detalles de la promoción
Sartorius™
YCW523-AC-00
200 g
140.00 €
Each
In Stock
to see stock.
Añadir a la cesta
 
11363304
Ver documentos Detalles de la promoción
Sartorius™
YCW553-AC-00
500 g
256.00 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Añadir a la cesta
 
11373304
Ver documentos Detalles de la promoción
Sartorius™
YCW613-AC-00
1 kg
281.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Añadir a la cesta
 
11383304
Ver documentos Detalles de la promoción
Sartorius™
YCW623-AC-00
2 kg
450.00 €
Each
In Stock
to see stock.
Añadir a la cesta
 
11393304
Ver documentos Detalles de la promoción
Sartorius™
YCW653-AC-00
5 kg
656.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Añadir a la cesta
 
11367494
Ver documentos Detalles de la promoción
Sartorius™
YCW713-AC-00
10 kg
1180.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Añadir a la cesta
 
Descripción

Descripción

  • Non-magnetizable
  • Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
  • Special Stainless steel construction with a density of 7,950 kg/m3
  • Complies with OIML R111-1 International Recommentation
Especificaciones

Especificaciones

Proof Line Weight
OIML Class F1
Special Stainless Steel
7,950kg/m3
Videos
SDS
Documentos

Documentos

Product Certifications