missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetyl chloride, 98%, Thermo Scientific™

Code produit. 15487767
Click to view available options
Quantity:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10166-09-3
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD03424714
InChI Key FKMIBYMTHOJWCY-UHFFFAOYSA-N
Synonym o-tolyl-acetyl chloride, o-tolylacetyl chloride, 2-2-methylphenyl acetyl chloride, o-methylphenylacetyl chloride, 2-methylphenylacetyl chloride, 2-methylbenzeneacetyl chloride, 2-methylphenyl acetyl chloride, 2-methylphenylacetic acid chloride
PubChem CID 2760632
IUPAC Name 2-(2-methylphenyl)acetyl chloride
SMILES CC1=CC=CC=C1CC(=O)Cl

Spécification

Color Pink
Boiling Point 84°C (1.5 mmHg)
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 168.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material o-Tolylacetyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'