missing translation for 'onlineSavingsMsg'
Learn More

o-Phenylenediamine, 98%, Thermo Scientific Chemicals

61.10 € - 382.91 €

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007721
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10542911
SDS View Documents
Thermo Scientific Acros
130552500
250 g Glass bottle
61.10 €
250g
Please to purchase this item. Need a web account? Register with us today!
10133420
SDS View Documents
Thermo Scientific Acros
130550010
1 kg Glass bottle
196.98 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10553504
SDS View Documents
Thermo Scientific Acros
130550050
5 kg Plastic drum
382.91 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
o-Phenylenediamine, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.