missing translation for 'onlineSavingsMsg'
Learn More

o-Anisoyl chloride, 97%, Thermo Scientific Chemicals

47.70 € - 167.00 €

Chemical Identifiers

CAS 21615-34-9
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00000664
InChI Key RZNHSEZOLFEFGB-UHFFFAOYSA-N
Synonym o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
PubChem CID 88969
IUPAC Name 2-methoxybenzoyl chloride
SMILES COC1=CC=CC=C1C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10124780
View Documents
Thermo Scientific Acros
276110050
5 g Glass bottle
47.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
10349440
View Documents
Thermo Scientific Acros
276110250
25 g Glass bottle
167.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21615-34-9
170.6
RZNHSEZOLFEFGB-UHFFFAOYSA-N
88969
COC1=CC=CC=C1C(=O)Cl
C8H7ClO2
MFCD00000664
o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
2-methoxybenzoyl chloride

Specifications

21615-34-9
100.0
1.1460g/mL
84°C
96% min. (GC)
CH3OC6H4COCl
MFCD00000664
1.146
Solubility in water: may decompose in water
COC1=CC=CC=C1C(=O)Cl
170.6
170.6
Liquid
96.0
Yellow to Brown
128°C to 129°C (8.0 mmHg)
Authentic
C8H7ClO2
1.5714 to 1.5734
10, 140
o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
RZNHSEZOLFEFGB-UHFFFAOYSA-N
2-methoxybenzoyl chloride
88969
97%
o-Anisoyl chloride, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 244-477-5

missing translation for 'tsca' : TSCA

Product Identifier
  • o-Anisoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.