missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Nylon Straight Threaded Coupling

Sturdy threaded fittings are designed for permanent installation. Use with rigid tubing for high-pressure applications

Brand:  Masterflex™ 06349-02

 View more versions of this product

Product Code. 11711084

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Straight threaded coupling
  • Available in packs of 6
  • Good chemical resistance
  • Rated to 200psi at 77°F (25°C)
  • Usable from -40 to 200°F (-40 to 93°C)
Specifications

Specifications

Nylon
6 Couplings
Ethylene Oxide
0.79 in.
Straight Threaded Coupling
3/8 in. NPT(F)
20 mm
Straight
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers