missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Nylon CellMicroSieves Square Sheets

Autoclavable and can be repeatedly washed and reused

102.00 € - 114.00 €

Specifications

Type Cell Micro Sieve
Material Nylon
For Use With (Application) Isolation of Specific Cell Types From a Dissociated Tissue Sample
Format Square Sheet
View More Specs

Products 2
Product Code Brand Pore Size Price Quantity & Availability  
Product Code Brand Pore Size Price Quantity & Availability  
12934257
View accessory products View Documents Promotion Details
Biodesign™
N35S
35 μm
102.00 €
Pack of 3
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
12984257
View accessory products View Documents Promotion Details
Biodesign™
N8S
8 μm
114.00 €
Pack of 3
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
Description

Description

Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory.

AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300 mL capacity are autoclavable and reusable.Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.

  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter
CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Specifications

Specifications

Cell Micro Sieve
Nylon
Isolation of Specific Cell Types From a Dissociated Tissue Sample
Square Sheet
Videos
SDS
Documents

Documents

Product Certifications