Learn More
Thermo Scientific™ Nunc™ 384 Well ShallowWell Standard Height Black and White Plates
Save costs in samples and reagents by reducing the total well volume to 25μL using Nunc 384-well shallow well standard height black and white polystyrene plates.
372.00 € - 959.00 €
Specifications
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
---|---|
Format | Standard |
Array | 16 x 24 |
Packaging | 100 Cases, 25 Pk. |
Product Line | Nunc |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10238343
|
Thermo Scientific™
264705 |
384 Shallow Well Standard Height, black, non-treated, no lid, packaged 25/100 |
372.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10689885
|
Thermo Scientific™
264706 |
384 Shallow Well Standard Height, white, non-treated, no lid, packaged 25/100 |
959.00 €
Pack of 100 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
Description
Accommodate small sample volumes by using Thermo Scientific™ Nunc™ 384-Well ShallowWell Standard Height Black and White Plates with total well volume of 25μL, ideal for high throughput screenings. The standard plate height allows for easy adaptation to most automation systems.
Features:- Black color minimizes back-scattered light and background noise
- ANSI™ (SBS) standard footprint
- Customized bar coding available
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
16 x 24 | |
Nunc | |
Polystyrene | |
384 | |
Flat Bottom (F-well) | |
25 |
Standard | |
100 Cases, 25 Pk. | |
Microplate | |
25 μL | |
2 to 20 μL | |
100 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.