missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey Nagel Bioanalysis™ Nucleospin Plasmid (Nolid)
Rapid Mini preparation of plasmid DNA on QIAcube
Brand: Macherey Nagel Bioanalysis™ 740499.240C
This item is not returnable.
View return policy
Description
240 preparations, Macherey NagelSpecifications
18°C to 25°C | |
Bacteria, E. coli | |
240 Preps | |
Plasmid DNA |
Mini Prep | |
High and low copy plasmid DNA purification | |
1−5 mL (high copy), 6−10 mL (low copy) |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %); guanidine hydrochloride; 2-propanol;Rnase