missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Plasmid (NoLid) Columns
Replacement columns for use with NucleoSpin™ Plasmid (NoLid) kits for rapid, small-scale preparation of high-purity, sequencing-grade plasmid DNA.
Brand: Macherey-Nagel™ 740499.50S
This item is not returnable.
View return policy
Description
NucleoSpin™ Plasmid (NoLid)
- For rapid small scale preparation high-purity plasmid DNA
- Fast procedure – 6 Mini preps in 25 min
- High capacity – up to 50 μg of plasmid DNA
- Chaotropic salt wash buffer – extra pure DNA
- Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
- High-copy plasmids from E. coli
- Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
- Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
- Supplementary protocol for M13 DNA availabl
Specifications
Clear | |
Silica Membrane Technology | |
Binding capacity: 50 μg | |
Sequencing-grade plasmid DNA | |
Preparation of high-purity plasmid DNA | |
NucleoSpin Plasmid (NoLid) Column | |
2 - 10 mL |
Mini Spin Column | |
E. coli Culture | |
Elution volume: 50 μL | |
NucleoSpin™ Plasmid (NoLid) Kit | |
Columns only | |
50 Each |
Safety and Handling
Product Identifier
Supplemental information
- NucleoSpin Plasmid (NoLid)
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %); guanidine hydrochloride; 2-propanol;Rnase