missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ DNA RapidLyse
Economical kit for the rapid isolation and purification of genomic DNA from tail and ear clippings, organs, and cells in one hour or less. Kits with 10, 50 or 250 preps available.
31.30 € - 585.00 €
Specifications
Format | Mini Spin Column |
---|---|
Isolation Technology | Silica Membrane Technology |
Sample Type | Eukaryotic Cells, Organs (Fresh, Frozen, Dried and Ethanol-Preserved), Tail and Ear Clippings |
Efficiency | Binding capacity: 60 μg |
Elution Volume | Elution volume: 60 - 100 μL |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15795412
|
Macherey-Nagel™
740100.10 |
10 |
31.30 €
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15715422
|
Macherey-Nagel™
740100.50 |
50 |
138.00 €
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15705422
|
Macherey-Nagel™
740100.250 |
250 |
585.00 €
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The isolation of genomic DNA (gDNA) can be a time-consuming task. The NucleoSpin™ DNA RapidLyse kit is designed with a unique buffer chemistry to minimize the expenditure of time, work, and money.
- Unique lysis chemistry to efficiently release gDNA from cells, tissues, and organs
- Powerful lysis in one hour or less
- Superior gDNA yields compared to standard extraction methods
- Highest cost-efficiency of gDNA purification products
Specifications
Mini Spin Column | |
Eukaryotic Cells, Organs (Fresh, Frozen, Dried and Ethanol-Preserved), Tail and Ear Clippings | |
Elution volume: 60 - 100 μL | |
Rapid extraction of genomic DNA | |
A260/A280: 1.7 - 1.9 | |
60 min or less |
Silica Membrane Technology | |
Binding capacity: 60 μg | |
genomic DNA | |
Columns, 2mL Collection tubes, Buffers, Poteinase K | |
Up to 40 mg fresh weight, 1 x 106 cells | |
Up to 4 μg DNA per mg tissue |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- NucleoSpin DNA RapidLyse
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : proteinase K, liquid(origin: tritirachium album)/1-dodecylpyridiniumchloride,guanidinium thiocyanat