missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ DNA Insect
Combines enzymatic lysis and mechanical disruption of cell walls for efficient isolation of genomic DNA from a wide variety of insects and crustacean samples (fresh, frozen, dried or ethanol preserved). Complete kit includes NucleoSpin™ Bead Tubes.
58.75 € - 221.00 €
Specifications
Content And Storage | Room temperature (18 - 25 °C) |
---|---|
Format | Mini Spin Column |
Isolation Technology | Silica MembraneTechnology and Mechanical (Bead Tube) Disruption |
Sample Type | Insect or Crustacean Specimens (Fresh, Frozen, Dried or Ethanol Preserved) |
Efficiency | Binding capacity: 60 μg |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15523626
|
Macherey-Nagel™
740470.10 |
10 |
58.75 €
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15533626
|
Macherey-Nagel™
740470.50 |
50 |
221.00 €
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isolation of genomic DNA from insects and crustaceans
- Fast and convenient procedure combining enzymatic lysis and bead disruption
- Suitable for any insect or crustacean (e.g., fruit fly, cricket, mealworm, midge larvae, mosquito)
- High quality DNA from fresh, frozen, dried or ethanol preserved specimen samples
- Bead Tubes included for difficult-to-lyse insects (due to exoskeletons and reinforced cell walls)
- Efficient lysis – Compatible with the most common disruption devices
- Typical downstream applications: multiplex PCR, Southern blotting, enzymatic reactions
Specifications
Room temperature (18 - 25 °C) | |
Silica MembraneTechnology and Mechanical (Bead Tube) Disruption | |
Binding capacity: 60 μg | |
Disruption device (e.g. Vortex-Genie 2 with MN Bead Tube Holder), Centrifuge, Vortex mixer | |
NucleoSpin™ DNA Insect Columns, NucleoSpin™ Bead Tubes Type D, Collection Tubes (2 mL), Buffers | |
< 40 mg | |
35 min/6 prep |
Mini Spin Column | |
Insect or Crustacean Specimens (Fresh, Frozen, Dried or Ethanol Preserved) | |
Elution volume: 25 - 200 μL | |
Isolation of genomic DNA from insects and crustaceans | |
A260/A280: 1.7 - 1.9 | |
Fragment size: 200 bp - approx. 50 kbp | |
< 25 μg (varies by sample and disruption device) |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year