missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Blood Column
Purify genomic DNA quickly and easily. NucleoSpin™ Blood Columns enable purification of genomic DNA from blood and biological fluids
45.45 € - 918.00 €
Specifications
Detection Method | Silica-membrane Technology |
---|---|
Format | Mini Spin Column |
Isolation Technology | Silica Membrane Technology |
For Use With (Equipment) | PCR, Southern Blotting, Enzymatic Reactions |
For Use With (Application) | For Genomic, Bacterial, Viral DNA Isolation |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11922512
|
Macherey-Nagel™
740951.10 |
10 |
45.45 €
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10559501
|
Macherey-Nagel™
740951.250 |
250 |
918.00 €
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11942402
|
Macherey-Nagel™
740951.50 |
50 |
188.00 €
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features- For rapid purification of high-quality DNA from blood
- Ready-to-use DNA in less than 30 min
- Complete removal of PCR inhibitors
- Suitable for whole blood, treated with citrate, EDTA, heparin, CPDA
- Consistently high DNA yields
- Suitable for isolation of viral DNA
- Genomic, bacterial, viral DNA isolation
- DNA from whole blood (human or animal blood, fresh or frozen)
- DNA from whole blood treated with citrate, EDTA, heparin, CPDA
- DNA from serum, plasma, buffy coat, platelets, body fluids (e.g. amniotic fluid)
- DNA from cultured cells
- DNA suitable for (PCR, Southern blotting, enzymatic reactions, etc.)
Specifications
Silica-membrane Technology | |
Silica Membrane Technology | |
For Genomic, Bacterial, Viral DNA Isolation | |
NucleoSpin™ Blood Column | |
30min. |
Mini Spin Column | |
PCR, Southern Blotting, Enzymatic Reactions | |
2mL Collection Tubes, Buffers, Proteinase K | |
1 Each | |
4 to 6μg |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash with water thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol