missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Bioanalysis™ Nucleospin 96 Gel And PCR Clean-Up Core Kit

Time saving medium to high throughput PCR clean up

Brand:  Macherey Nagel Bioanalysis™ 740464.4


Product Code. 12718432

  • 744.00 € / 384 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

4 x 96 preps for the purification of PCR products, buffers, NucleoSpin™ Extract binding plate, user manual (formerly know as NucleoSpin™ 96 Extract II) Macherey-Nagel
Specifications

Specifications

18°C to 25°C
PCR Reaction Mixture
384 Preps
50 bp−approx. 10 kbp
96-well Plate
PCR clean up
< 100 L
Clean up
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers