Learn More
Invitrogen™ NT-3/NTF3 Human ELISA Kit
Sandwich ELISA Kit
668.00 € - 2635.00 €
Specifications
Accession Number | P20783 |
---|---|
Assay Sensitivity | 4 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15405093
|
Invitrogen™
EHNTF3 |
96 Tests |
668.00 €
1 set |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
17135529
|
Invitrogen™
EHNTF3X5 |
5 x 96 Tests |
2635.00 €
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
Description
The Human NeuroTrophic Factor 3 (NT-3) (Hu NTF3) ELISA quantitates Hu NTF3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NTF3. Principle of the method The Human NTF3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.Specifications
P20783 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
HDNF,NGF-2,NGF2,NT-3,NT3 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
4 pg/mL | |
Biotin | |
Kinase & Inhibitors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
4908 | |
<12% | |
HRP | |
Neurobiology | |
HDNF, NGF-2, NGF2, NT-3, NT3 | |
1 hr. 20 min. |