Learn More
NR1D2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13906-1-AP-150UL
 View more versions of this product
Product Code. 16882324
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
Specifications
Specifications
NR1D2
Polyclonal
Unconjugated
NR1D2
BD73, EAR 1R, NR1D2, Rev erb beta, Reverb Beta, RVR
Rabbit
Antigen Affinity Chromatography
RUO
259241, 353187, 9975
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14995, Q60674, Q63504
Nr1d2
NR1D2 Fusion Protein Ag4881
150 μL
Primary
Human, Mouse, Rat