Learn More
NPR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55116-1-AP-150UL
 View more versions of this product
Product Code. 16806575
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Guanylyl cyclases, catalyzing the production of cGMP from GTP, are classified as soluble and membrane forms. The membrane guanylyl cyclases, often termed guanylyl cyclases A through F, form a family of cell-surface receptors with a similar topographic structure: an extracellular ligand-binding domain, a single membrane-spanning domain, and an intracellular region that contains a protein kinase-like domain and a cyclase catalytic domain. GC-A and GC-B function as receptors for natriuretic peptides; they are also referred to as atrial natriuretic peptide receptor A and type B. Also see NPR3, which encodes a protein with only the ligand-binding transmembrane and 37-amino acid cytoplasmic domains. NPR1 is a membrane-bound guanylate cyclase that serves as the receptor for both atrial and brain natriuretic peptides and BNP, respectively).
Specifications
Specifications
NPR1
Polyclonal
Unconjugated
NPR1
ANP A, ANPa, ANPR A, ANPRA, GC A, Guanylate cyclase A, GUC2A, GUCY2A, NPR1
Rabbit
Antigen Affinity Chromatography
RUO
4881
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
P16066
NPR1
Peptide
150 μL