Learn More
Invitrogen™ Novex™ IL10 Mouse Antibody Pair
Concentrated
Brand: Invitrogen™ CMC0103
Includes: Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet
Requires : Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).
Description
Antibody Pair Kit comes with matched antibody pairs to detect and quantify protein levels of mouse IL-10.
Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate.
Build Your Own Immunoassay, Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice
Specifications
NM_010548.2, NP_034678.1 | |
CSIF, Il-10 | |
Il10 |