Learn More
Invitrogen™ IL-18 Rat ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ KRC2341
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Rat Interleukin 18 (Rt IL-18) ELISA quantitates Rt IL-18 in rat serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt IL-18. Principle of the method The Rat IL-18 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.Order Info
Shipping Condition: Room Temperature
Specifications
P97636 | |
<4 pg/mL | |
ELISA Kit | |
Rat | |
Colorimetric Microplate Reader | |
Il18 | |
4.9% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
IL-18 | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
15.6-1000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric Detection | |
29197 | |
4 hr. | |
3.9% | |
HRP | |
96 Tests | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Rat | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.