missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTCH1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20687-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to NOTCH1. It can recognize the full length NOTCH1 (270 kDa) and all the three cleaved NOTCH1 forms.
This gene is similar in sequence to the S. cerevisiae YDL201w gene. The gene product contains a conserved S-adenosylmethionine-binding motif and is inactivated by phosphorylation.Specifications
NOTCH1 | |
Polyclonal | |
Unconjugated | |
NOTCH1 | |
hN1, Notch 1, NOTCH1, TAN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4851 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P46531 | |
NOTCH1 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |