missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Normal Goat Serum (10%)
Normal Goat Serum (10%)
Brand: Invitrogen 50062Z
This item is not returnable.
View return policy
Description
This 10% Normal Goat Serum blocking solution is ready-to-use, in phosphate buffered saline (pH 7.4) with 0.1% sodium azide. This normal goat serum is intended for use as a blocking solution when a second antibody of goat origin is applied in an immunohistochemical staining procedure.To block non-specific binding from the goat second antibody, incubate the tissue section with 10% Normal Goat Serum for 10 minutes at room temperature prior to the primary antibody incubation step.
Specifications
Store at 2°C to 8°C. Do not freeze. | |
Goat/Caprine | |
100 mL | |
Wet Ice |
For Laboratory Use