missing translation for 'onlineSavingsMsg'
Learn More

Nonafluorobutanesulfonyl fluoride, 90+%, Thermo Scientific Chemicals

28.14 € - 226.13 €

Chemical Identifiers

CAS 375-72-4
Molecular Formula C4F10O2S
Molecular Weight (g/mol) 302.086
MDL Number MFCD00007422
InChI Key LUYQYZLEHLTPBH-UHFFFAOYSA-N
Synonym nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro
PubChem CID 67814
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride
SMILES C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418690
View Documents
Thermo Scientific Alfa Aesar
B21322.06
5 g
28.14 €
5g
Please to purchase this item. Need a web account? Register with us today!
11439237
View Documents
Thermo Scientific Alfa Aesar
B21322.14
25 g
79.50 €
25g
Please to purchase this item. Need a web account? Register with us today!
11449237
View Documents
Thermo Scientific Alfa Aesar
B21322.22
100 g
226.13 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Benzynes were generated from o-(trimethylsilyl) phenols using nonafluorobutanesulfonyl fluoride (NfF) by a domino process.Nonafluorobutanesulfonyl fluoride (nonaflyl fluoride, C 4 F 9 SO 2 F, NfF) is the most widely used commercially available reagent for the synthesis of nonaflates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzynes were generated from o-(trimethylsilyl) phenols using nonafluorobutanesulfonyl fluoride (NfF) by a domino process.Nonafluorobutanesulfonyl fluoride (nonaflyl fluoride, C 4 F 9 SO 2 F, NfF) is the most widely used commercially available reagent for the synthesis of nonaflates.

Solubility
Hydrolyses in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, baese..
Specifications

Chemical Identifiers

375-72-4
302.086
LUYQYZLEHLTPBH-UHFFFAOYSA-N
67814
C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F
C4F10O2S
MFCD00007422
nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro
1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride

Specifications

375-72-4
1.75
None
1.281
UN3265
Moisture sensitive
Hydrolyses in water.
C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F
302.086
302.09
Nonafluorobutanesulfonyl fluoride
-110°C
65°C to 66°C
C4F10O2S
MFCD00007422
1813589
nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro
LUYQYZLEHLTPBH-UHFFFAOYSA-N
1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride
67814
≥90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335-H370

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 206-792-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only