missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIAMRE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13888-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determit of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
NKIAMRE | |
Polyclonal | |
Unconjugated | |
CDKL3 | |
CDKL3, cyclin dependent kinase like 3, NKIAMRE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
213084, 51265, 60396 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BLF2, Q8IVW4, Q9JM01 | |
CDKL3 | |
NKIAMRE Fusion Protein Ag4812 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction