missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13884-1-AP-20UL
This item is not returnable.
View return policy
Description
NKCC1 mediates sodium and chloride transport and reabsorption. NKCC1 is a membrane protein and is important in maintaining proper ionic balance and cell volume. NKCC1 is an electrically silent transporter system that mediates sodium and chloride reabsorption. Further, NKCC1 plays a vital role in the regulation of ionic balance and cell volume and is phosphorylated in response to DNA damage. Three transcript variants encoding two different isoforms have been found for NKCC1. Diseases associated with SLC12A2 include Basosquamous Carcinoma and Metatypical Basal Cell Carcinoma.Specifications
NKCC1 | |
Polyclonal | |
Unconjugated | |
SLC12A2 | |
Basolateral Na K Cl symporter, BSC, BSC2, NKCC1, SLC12A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20496, 6558, 83629 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55011, P55012 | |
SLC12A2 | |
NKCC1 Fusion Protein Ag4991 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction