Learn More
Nitromethane-d3, for NMR, 99 atom % D, Thermo Scientific Chemicals
69.40 € - 226.00 €
Chemical Identifiers
CAS | 13031-32-8 |
---|---|
Molecular Formula | CH3NO2 |
Molecular Weight (g/mol) | 64.06 |
MDL Number | MFCD00044214 |
InChI Key | LYGJENNIWJXYER-FIBGUPNXSA-N |
Synonym | nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms |
PubChem CID | 123293 |
IUPAC Name | trideuterio(nitro)methane |
SMILES | [2H]C([2H])([2H])[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10104230
|
Thermo Scientific
166370050 |
5 mL | Glass Bottle |
69.40 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10533531
|
Thermo Scientific Acros
166370100 |
10 mL | Glass Bottle |
123.00 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772011
|
Thermo Scientific Acros
166370250 |
25 mL | Glass Bottle |
226.00 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
13031-32-8 | |
64.06 | |
LYGJENNIWJXYER-FIBGUPNXSA-N | |
123293 | |
[2H]C([2H])([2H])[N+]([O-])=O |
CH3NO2 | |
MFCD00044214 | |
nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms | |
trideuterio(nitro)methane |
Specifications
Liquid | |
1.1800g/mL | |
35°C | |
1.18 | |
13031-32-8 | |
CH3NO2 | |
MFCD00044214 | |
LYGJENNIWJXYER-FIBGUPNXSA-N | |
trideuterio(nitro)methane | |
123293 | |
NMR |
Colorless | |
98.5% min. | |
1.3795 | |
0.1% max. (K.F.) | |
100.0°C | |
CD3NO2 | |
nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms | |
[2H]C([2H])([2H])[N+]([O-])=O | |
64.06 | |
64.06 | |
Nitromethane-d3, 99 atom % D |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 235-892-2
- Nitromethane-d3
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
- Flammable liquid Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only