missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane, 99%, pure, Thermo Scientific Chemicals

267.33 € - 902.49 €

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10625582
View Documents
Thermo Scientific Acros
148510025
2.5 L Glass Bottle
267.33 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11309811
View Documents
Thermo Scientific Acros
148510100
10 L Metal drum
902.49 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-52-5
61.04
LYGJENNIWJXYER-UHFFFAOYSA-N
6375
nitromethane
CH3NO2
MFCD00007400
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
CHEBI:77701
C[N+]([O-])=O

Specifications

75-52-5
1.1200g/mL
35°C
98.5% min. (GC)
CH3NO2
MFCD00007400
01,739; 04,357; 09,323; 13,199
1.12
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
C[N+]([O-])=O
61.04
CHEBI:77701
61.04
Pure
Nitromethane
-29.0°C
100.0°C to 102.0°C
Authentic
CH3NO2
1.3805 to 1.3825
01,74
15,6698
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
LYGJENNIWJXYER-UHFFFAOYSA-N
nitromethane
6375
0.61 mPa.s (25°C)
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-876-6

Product Identifier
  • Nitromethane

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.