missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Nitritex™ Omega Pro-tech™ Blue Disposable Powder-free Laboratory Gloves

Product Code. 12920845
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Blue disposable powder free gloves resist cuts and snags and are available in a variety of sizes.

  • Blue
  • Beaded cuff
  • Non-sterile
  • Nitrile contains no proteins
  • Various sizes
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 2859
Classification Category 3 - Complex Design Personal Protective Equipment
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Product Type Nitrile Examination Glove
Size Large
Glove Cuff Style Beaded/Rolled
Inspected for Medical Use Yes
Length (Metric) 24 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'