missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, TraceSELECT™, for trace analysis, ≥69.0%, Honeywell Fluka™

Product Code. 15607300
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
5 L
6 x 500 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

TraceSELECT™, for trace analysis,≥69.0%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2, 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Boiling Point 122°C
CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Density 1.41 g/mL at 20 °C
Vapor Pressure 8 mmHg (20°C)
Quantity 5 L
Linear Formula HNO3
UN Number UN2031
Formula Weight 63.01
Percent Purity ≥69.0% (T)
Chemical Name or Material Nitric acid, TraceSELECT™, for trace analysis, ≥69.0%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitric acid
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'