missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, Environmental Grade, 67-70%, Thermo Scientific Chemicals

156.78 €

Chemical Identifiers

CAS 7697-37-2, 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471497
View Documents
Thermo Scientific Alfa Aesar
038747.AP
500 mL
156.78 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Main commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7697-37-2, 7732-18-5
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O

Specifications

Colorless to pale yellow liquid
7697-37-2, 7732-18-5
1.41
23 hPa (17mm Hg) at 20°C
HNO3
MFCD00011349
Miscible in water and ether.
O[N+]([O-])=O
63.01
CHEBI:48107
Environmental
Nitric acid
Colorless to Yellow
121°C
Pungent
67-70%
Liquid,single sub-boiling quartz distillation
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01
Nitric acid: 69.0%; Water: 31.0%
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c

H272-H290-H314-H331

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II

missing translation for 'einecsNumber' : 231-714-2

missing translation for 'rtecsNumber' : QU5775000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Nitric acid, 67-70%

Signal Word
  • Danger

Hazard Category
  • Oxidising liquid Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P320-Specific treatment is urgent (see on this label).
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Nitric acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.