missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, ACS Reagent, 70%, Honeywell Fluka™
ACS Reagent, 70%
63.87 € - 785.07 €
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15685520
|
Honeywell-Fluka
438073-100ML |
100 mL | Glass Bottle |
63.87 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15615530
|
Honeywell-Fluka
438073-500ML |
500 mL | Glass Bottle |
200.12 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15695520
|
Honeywell-Fluka
438073-2.2L-P |
2.2 L | Poly Bottle |
120.22 €
2.20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15605530
|
Honeywell-Fluka
438073-2.5L |
2.5 L | Glass Bottle |
341.84 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15625530
|
Honeywell-Fluka
438073-6X500ML |
6 x 500 mL | Glass Bottle | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
Acidic | |
-38°C | |
1.413g/mL at 20°C | |
8mmHg (20°C) | |
Liquid | |
UN2031 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01g/mol | |
Nitric acid |
7697-37-2 | |
122°C (1013hPa) | |
Stinging | |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
68.0 to 70.0% |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
missing translation for 'dotInformation' : class 8 (5.1) - PG 2 - IATA PAX forbidden - Nitric acid ≥65% - ≤70%
missing translation for 'rtecsNumber' : 231-714-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Nitric acid
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title