Learn More
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals
20.40 € - 79.60 €
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341578
|
Thermo Scientific Alfa Aesar
010984.AC |
25 mL |
20.40 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331578
|
Thermo Scientific Alfa Aesar
010984.AK |
250 mL |
54.10 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395809
|
Thermo Scientific Alfa Aesar
010984.AP |
500 mL |
79.60 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nitric acid, 65-70% is used in as a nitrating or an oxidizing agent in organic synthesis. It is industrially used for the manufacturing of fertilizers. Further, it finds use in manufacture of explosives, nylon precursors, specialty organic compounds and drug intermediates.It is also used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple. It is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
Colorless to Yellow | |
121°C | |
1.41 g/mL | |
HNO3 | |
MFCD00011349 | |
14,6577 | |
Miscible with water. | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
99.9999% | |
Nitric acid |
7697-37-2 | |
65 to 70% | |
Pungent | |
Liquid | |
UN2031 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.
P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H331
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID
missing translation for 'einecsNumber' : 231-714-2
missing translation for 'rtecsNumber' : QU5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only