missing translation for 'onlineSavingsMsg'
Learn More

Nitrate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12982155
250 mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 14797-55-8
InChI Key NHNBFGGVMKEFGY-UHFFFAOYSA-N
PubChem CID 943
ChEBI CHEBI:17632
IUPAC Name nitrate

Specifications

Packaging Bottle
Quantity 250 mL
Molecular Formula NO3-
Physical Form Liquid
Concentration 1 g/L
SMILES [N+](=O)([O-])[O-]
Molecular Weight (g/mol) 62.004
Type Single-component Standard Solution
Solvent or Matrix H2O
Chemical Name or Material Nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitrate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.