Learn More
Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
37.39 € - 846.21 €
Chemical Identifiers
CAS | 7440-03-1 |
---|---|
Molecular Formula | Nb |
Molecular Weight (g/mol) | 92.91 |
MDL Number | MFCD00011126 |
InChI Key | GUCVJGMIXFAOAE-UHFFFAOYSA-N |
Synonym | columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio |
PubChem CID | 23936 |
ChEBI | CHEBI:33344 |
IUPAC Name | niobium |
SMILES | [Nb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465935
|
Thermo Scientific Alfa Aesar
010275.88 |
2.5 g |
37.39 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311348
|
Thermo Scientific Alfa Aesar
010275.14 |
25 g |
85.83 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391338
|
Thermo Scientific Alfa Aesar
010275.36 |
500 g |
846.21 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-03-1 | |
92.91 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
23936 | |
niobium |
Nb | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
CHEBI:33344 | |
[Nb] |
Specifications
7440-03-1 | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
[Nb] | |
92.91 | |
CHEBI:33344 | |
−325 Mesh | |
Powder | |
Niobium powder |
Nb | |
UN3089 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
niobium | |
23936 | |
99.8% | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 231-113-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only