Learn More
Cole-Parmer™ Nine-Position Stirring Hot Plate
Nine-Position Stirring Hot Plate offers more versatility than single-position units. Each stirring position is individually controlled so you can perform different mixing applications simultaneously.
Brand:  Cole-Parmer™ 51450-73
Product Code. 15560492
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Each stirring position is individually controlled to perform different mixing applications at the same time
    • Front panel controls are positioned to prevent you from touching the hot plate
Specifications
Specifications
Ceramic
100 to 1500 rpm
17.5 in.
100°C to 450°C
4.5 in.
30.48 cm
44.45 cm
Stirring Hotplate
Multiple
120 V
6 ft. Cord with Plug
11.43 cm
212°F to 845°F
60 Hz
12 amp
12 in.
40 lb.
0.4 L