Learn More
Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm, Thermo Scientific Chemicals
160.00 € - 578.00 €
Chemical Identifiers
CAS | 14177-51-6 |
---|---|
Molecular Formula | NiO4W |
Molecular Weight (g/mol) | 306.529 |
MDL Number | MFCD00016268 |
InChI Key | QLTKZXWDJGMCAR-UHFFFAOYSA-N |
Synonym | nickel tungstate, nickel tungsten tetraoxide, nickel tungsten oxide niwo4, nickel 2+ dioxido dioxo tungsten, nickel 2+ ion tungstate, dioxido dioxo tungsten; nickel 2+ |
PubChem CID | 84240 |
IUPAC Name | dioxido(dioxo)tungsten;nickel(2+) |
SMILES | [O-][W](=O)(=O)[O-].[Ni+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384777
|
Thermo Scientific Alfa Aesar
041826.14 |
25 g |
160.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394777
|
Thermo Scientific Alfa Aesar
041826.22 |
100 g |
578.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel tungsten oxide is used as photo catalysts, sensors and humidity dielectric resonators. It is considered as a new cathode material for asymmetric super capacitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14177-51-6 | |
306.529 | |
QLTKZXWDJGMCAR-UHFFFAOYSA-N | |
84240 | |
[O-][W](=O)(=O)[O-].[Ni+2] |
NiO4W | |
MFCD00016268 | |
nickel tungstate, nickel tungsten tetraoxide, nickel tungsten oxide niwo4, nickel 2+ dioxido dioxo tungsten, nickel 2+ ion tungstate, dioxido dioxo tungsten; nickel 2+ | |
dioxido(dioxo)tungsten;nickel(2+) |
Specifications
14177-51-6 | |
NiO4W | |
nickel tungstate, nickel tungsten tetraoxide, nickel tungsten oxide niwo4, nickel 2+ dioxido dioxo tungsten, nickel 2+ ion tungstate, dioxido dioxo tungsten; nickel 2+ | |
QLTKZXWDJGMCAR-UHFFFAOYSA-N | |
dioxido(dioxo)tungsten;nickel(2+) | |
84240 | |
99.9% | |
(metals basis excluding Co), Co <100ppm |
Powder | |
MFCD00016268 | |
Insoluble in water. | |
[O-][W](=O)(=O)[O-].[Ni+2] | |
306.529 | |
306.53 | |
Odorless | |
Nickel tungsten oxide |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
missing translation for 'einecsNumber' : 238-032-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only