missing translation for 'onlineSavingsMsg'
Learn More

Nickel tubing, 0.81mm (0.032in) OD, 0.51mm (0.02in) ID, 99% (metals basis), Thermo Scientific™

Product Code. 11326828
Click to view available options
Quantity:
60 cm
5 x 60 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 5 x 60 cm
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Nickel tubing, OD: 0.81mm (0.032 in.) ID: 0.51mm (0.02 in.)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'