missing translation for 'onlineSavingsMsg'
Learn More

Nickel powder, spherical, -300 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

29.25 € - 511.55 €

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11340739
SDS View Documents
Thermo Scientific Alfa Aesar
010581.09
10 g
29.25 €
10g
Please to purchase this item. Need a web account? Register with us today!
11360739
SDS View Documents
Thermo Scientific Alfa Aesar
010581.22
100 g
60.50 €
100g
Please to purchase this item. Need a web account? Register with us today!
11330739
SDS View Documents
Thermo Scientific Alfa Aesar
010581.36
500 g
156.78 €
500g
Please to purchase this item. Need a web account? Register with us today!
11350739
SDS View Documents
Thermo Scientific Alfa Aesar
010581.A3
2 kg
511.55 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H228-H317-H350-H372

missing translation for 'dotInformation' : DOT Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-111-4

missing translation for 'rtecsNumber' : QR5950000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel powder, spherical, -300 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.