Learn More
Nickel molybdenum oxide, 98%, Thermo Scientific Chemicals
55.17 € - 252.26 €
Chemical Identifiers
CAS | 14177-55-0 |
---|---|
Molecular Formula | MoNiO4 |
Molecular Weight (g/mol) | 218.639 |
MDL Number | MFCD00016255 |
InChI Key | NLPVCCRZRNXTLT-UHFFFAOYSA-N |
Synonym | molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+ |
PubChem CID | 84241 |
IUPAC Name | dioxido(dioxo)molybdenum;nickel(2+) |
SMILES | [O-][Mo](=O)(=O)[O-].[Ni+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443870
|
Thermo Scientific Alfa Aesar
089938.14 |
25 g |
55.17 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433870
|
Thermo Scientific Alfa Aesar
089938.22 |
100 g |
252.26 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel molybdenum oxide is used as catalyst in petroleum industry for the oil fraction like naptha, kerosene and diesel. It is also used extensively as a chemical intermediate for catalytic purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14177-55-0 | |
218.639 | |
NLPVCCRZRNXTLT-UHFFFAOYSA-N | |
84241 | |
[O-][Mo](=O)(=O)[O-].[Ni+2] |
MoNiO4 | |
MFCD00016255 | |
molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+ | |
dioxido(dioxo)molybdenum;nickel(2+) |
Specifications
14177-55-0 | |
MoNiO4 | |
molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+ | |
NLPVCCRZRNXTLT-UHFFFAOYSA-N | |
dioxido(dioxo)molybdenum;nickel(2+) | |
84241 | |
98% | |
Nickel molybdenum oxide |
Powder | |
MFCD00016255 | |
Soluble in water (4652mg/L at 20C). | |
[O-][Mo](=O)(=O)[O-].[Ni+2] | |
218.639 | |
218.65 | |
Odorless |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
missing translation for 'einecsNumber' : 238-034-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.