Learn More
Nickel(II) tetrafluoroborate hexahydrate, 99%, Thermo Scientific Chemicals
30.50 € - 40.00 €
Chemical Identifiers
CAS | 15684-36-3, 14708-14-6 |
---|---|
Molecular Formula | B2F8H12NiO6 |
Molecular Weight (g/mol) | 340.39 |
MDL Number | MFCD00150259 |
InChI Key | BFFWGFMAXHCLDV-UHFFFAOYSA-N |
Synonym | nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate |
PubChem CID | 177615 |
SMILES | O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10621073
|
Thermo Scientific Acros
358730100 |
10 g | Glass bottle |
30.50 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10405862
|
Thermo Scientific Acros
358730250 |
25 g | Glass bottle |
40.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15684-36-3, 14708-14-6 | |
340.39 | |
BFFWGFMAXHCLDV-UHFFFAOYSA-N | |
177615 |
B2F8H12NiO6 | |
MFCD00150259 | |
nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate | |
O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F |
Specifications
15684-36-3, 14708-14-6 | |
98.5 | |
Crystals | |
B2F8H12NiO6 | |
MFCD00150259 | |
Solubility in water: soluble in water | |
O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F | |
340.39 | |
340.39 | |
Nickel(II) tetrafluoroborate hexahydrate |
Blue-Green | |
100.0 | |
98.5% min. (Complexometry) | |
Ni(BF4)2·6H2O | |
nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate | |
BFFWGFMAXHCLDV-UHFFFAOYSA-N | |
nickel(2+) bis(tetrafluoroboranuide) hexahydrate | |
177615 | |
99% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause cancer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 238-753-4
missing translation for 'rtecsNumber' : QR6650000
missing translation for 'tsca' : TSCA
- Nickel(II) tetrafluoroborate hexahydrate
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only