Learn More
Nickel(II) oxide, green, Ni 78.5%, Thermo Scientific Chemicals
104.52 € - 325.62 €
Chemical Identifiers
CAS | 1313-99-1 |
---|---|
Molecular Formula | NiO |
Molecular Weight (g/mol) | 74.69 |
MDL Number | MFCD00011145 |
InChI Key | GNRSAWUEBMWBQH-UHFFFAOYSA-N |
Synonym | nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide |
PubChem CID | 14805 |
IUPAC Name | oxonickel |
SMILES | O=[Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402157
|
Thermo Scientific Alfa Aesar
045094.30 |
250 g |
104.52 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412157
|
Thermo Scientific Alfa Aesar
045094.A1 |
1 kg |
325.62 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel(II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs). It is blended with other high purity oxides and used in a variety of semiconductor applications such as thermistors and varistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-99-1 | |
74.69 | |
GNRSAWUEBMWBQH-UHFFFAOYSA-N | |
14805 | |
O=[Ni] |
NiO | |
MFCD00011145 | |
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide | |
oxonickel |
Specifications
1984°C | |
Green | |
Ni 78.5% | |
MFCD00011145 | |
Soluble in ammonium hydroxide. Insoluble in water. | |
O=[Ni] | |
74.69 | |
74.71 | |
6.67 |
1313-99-1 | |
Powder | |
NiO | |
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide | |
GNRSAWUEBMWBQH-UHFFFAOYSA-N | |
oxonickel | |
14805 | |
Odorless | |
Nickel(II) oxide |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
missing translation for 'einecsNumber' : 215-215-7
missing translation for 'rtecsNumber' : QR8400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel(II) oxide, green
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
- Carcinogenicity Category 1A
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H350i-May cause cancer by inhalation.
- H372-Causes damage to organs.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : Nickel oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.