Learn More
Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals
24.52 € - 396.98 €
Chemical Identifiers
CAS | 13478-00-7 |
---|---|
Molecular Formula | H12N2NiO12 |
Molecular Weight (g/mol) | 290.79 |
MDL Number | MFCD00149805 |
InChI Key | AOPCKOPZYFFEDA-UHFFFAOYSA-N |
Synonym | nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 |
PubChem CID | 61630 |
SMILES | O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408740
|
Thermo Scientific Alfa Aesar
A15540.22 |
100 g |
24.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418740
|
Thermo Scientific Alfa Aesar
A15540.36 |
500 g |
56.18 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11482967
|
Thermo Scientific Alfa Aesar
A15540.0B |
1000 g |
101.51 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492967
|
Thermo Scientific Alfa Aesar
A15540.0I |
5000 g |
396.98 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428740
|
Thermo Scientific Alfa Aesar
A15540.A4 |
2.5 kg |
223.11 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.
Solubility
Very soluble in water. Soluble in ethanol.
Notes
As an oxidizer, it is incompatible with reducing agents.
Chemical Identifiers
13478-00-7 | |
290.79 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
61630 |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
137°C | |
13478-00-7 | |
Ni(NO3)2·6H2O | |
UN2725 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+);dinitrate;hexahydrate | |
61630 | |
98% | |
Hygroscopic | |
Nickel(II) nitrate hexahydrate |
56.7°C | |
H12N2NiO12 | |
MFCD00149805 | |
14,6513 | |
Very soluble in water. Soluble in ethanol. | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
290.79 | |
290.80 (182.70 Anhydrous) | |
Odorless | |
2.05 |
Safety and Handling
GHS H Statement
H272-H334-H350-H360-H372-H341-H318-H302-H332-H315-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
H272-H302+H332-H315-H317-H318-H334-H335-H341-H350i-H360D-H372
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NICKEL NITRATE
missing translation for 'einecsNumber' : 236-068-5
missing translation for 'rtecsNumber' : QR7300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel(II) nitrate hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Nickel(II) nitrate hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.