missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) lactate tetrahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11490038
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H10NiO6·4H2O
MDL Number MFCD02683106

Specifications

Quantity 500 g
UN Number UN3077
Formula Weight 308.91 (236.85 Anhydrous)
Percent Purity 98%
Chemical Name or Material Nickel(II) lactate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.