missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) lactate tetrahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11480038
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H10NiO6·4H2O
MDL Number MFCD02683106

Specifications

Quantity 100 g
UN Number UN3077
Formula Weight 308.91 (236.85 Anhydrous)
Percent Purity 98%
Chemical Name or Material Nickel(II) lactate tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'