missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) hydroxide, Ni 61%, Thermo Scientific Chemicals

Product Code. 15461017
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Due to its redox behavior, it finds use in rechargeable battery electrodes. It is useful for the storage of electrochemical energy, with potential applications in car batteries. It is a potential material for several biological applications, for example in chromosomal DNA quantification assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-48-7
Molecular Formula H2NiO2
Molecular Weight (g/mol) 92.71
MDL Number MFCD00011140
InChI Key BFDHFSHZJLFAMC-UHFFFAOYSA-L
IUPAC Name nickel(2+) dihydroxide
SMILES [OH-].[OH-].[Ni++]

Specifications

Melting Point 230°C (decomposition)
Color Green
Physical Form Powder
Quantity 5 kg
Linear Formula Ni(OH)2
UN Number UN3077
Formula Weight 92.7
Concentration or Composition (by Analyte or Components) Ni 61%
Odor Odorless
Density 4.1g/mL
Chemical Name or Material Nickel(II) hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'