missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) hydroxide, Ni 61%, Thermo Scientific Chemicals

Product Code. 11301849
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Due to its redox behavior, it finds use in rechargeable battery electrodes. It is useful for the storage of electrochemical energy, with potential applications in car batteries. It is a potential material for several biological applications, for example in chromosomal DNA quantification assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-48-7
Molecular Formula H2NiO2
Molecular Weight (g/mol) 92.71
MDL Number MFCD00011140
InChI Key BFDHFSHZJLFAMC-UHFFFAOYSA-L
IUPAC Name nickel(2+) dihydroxide
SMILES [OH-].[OH-].[Ni++]

Specifications

Melting Point 230°C (decomposition)
Color Green
Physical Form Powder
Quantity 5 g
Linear Formula Ni(OH)2
UN Number UN3077
Formula Weight 92.7
Concentration or Composition (by Analyte or Components) Ni 61%
Odor Odorless
Density 4.1g/mL
Chemical Name or Material Nickel(II) hydroxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.