Learn More
Nickel(II) cyanide tetrahydrate, Thermo Scientific Chemicals
For Metallurgy, Nickel-Plating
153.00 € - 729.00 €
Chemical Identifiers
CAS | 13477-95-7 |
---|---|
Molecular Formula | C2H8N2NiO4 |
Molecular Weight (g/mol) | 182.789 |
MDL Number | MFCD00049490 |
InChI Key | VHHVAACTKRUWEK-UHFFFAOYSA-N |
Synonym | nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate |
PubChem CID | 71317429 |
IUPAC Name | nickel(2+);dicyanide;tetrahydrate |
SMILES | [C-]#N.[C-]#N.O.O.O.O.[Ni+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378278
|
Thermo Scientific Alfa Aesar
039480.09 |
10 g |
153.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388278
|
Thermo Scientific Alfa Aesar
039480.18 |
50 g |
729.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Applied in metallurgy, nickel-plating industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13477-95-7 | |
182.789 | |
VHHVAACTKRUWEK-UHFFFAOYSA-N | |
71317429 | |
[C-]#N.[C-]#N.O.O.O.O.[Ni+2] |
C2H8N2NiO4 | |
MFCD00049490 | |
nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate | |
nickel(2+);dicyanide;tetrahydrate |
Specifications
200°C-4H2O | |
Green | |
C2H8N2NiO4 | |
nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate | |
VHHVAACTKRUWEK-UHFFFAOYSA-N | |
nickel(2+);dicyanide;tetrahydrate | |
71317429 | |
Light green powder |
13477-95-7 | |
Powder | |
MFCD00049490 | |
Insoluble in water. Soluble in ammonium carbonate,ammonia,alkali cyanides | |
[C-]#N.[C-]#N.O.O.O.O.[Ni+2] | |
182.789 | |
182.73 (110.73 Anhydrous) | |
Nickel(II) cyanide tetrahydrate |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350-H372
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II
missing translation for 'einecsNumber' : 209-160-8
missing translation for 'rtecsNumber' : QR6495000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only