missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) chloride hexahydrate, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
Melting Point | 140°C-H2O |
Color | Green |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | 99.95% (metals basis) |
Solubility Information | Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide. |
Formula Weight | 237.71 (129.60 Anhydrous) |
Odor | Odorless |
Density | 1.92 |
Chemical Name or Material | Nickel(II) chloride hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'