missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 97%, Thermo Scientific Chemicals

40.70 € - 256.28 €

Chemical Identifiers

CAS 7791-20-0, 7718-54-9
Molecular Formula Cl2Ni·6H2O
Molecular Weight (g/mol) 237.71
MDL Number MFCD00149809
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10471631
SDS View Documents
Thermo Scientific Acros
207670250
25 g Glass bottle
40.70 €
25g
Please to purchase this item. Need a web account? Register with us today!
10368390
SDS View Documents
Thermo Scientific Acros
207675000
500 g Glass bottle
67.13 €
500g
Please to purchase this item. Need a web account? Register with us today!
10582031
SDS View Documents
Thermo Scientific Acros
207670020
2 kg Glass bottle
256.28 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-20-0, 7718-54-9
Cl2Ni·6H2O
237.71
MFCD00149809

Specifications

7791-20-0, 7718-54-9
96.0
4 to 6 (5% soln. at 20°C)
96% min. (Complexometry)
NiCl2·6H2O
14,351; 15,96; 17,86
Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
237.71
Nickel(II) chloride hexahydrate, 97%
Green
100.0
Crystalline Powder and/or Crystals
Cl2Ni·6H2O
MFCD00149809
15, 6590
237.71
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid relea

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : QR6480000

Product Identifier
  • Nickel(II) chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Nickel(II) chloridehexahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.