missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, 99.99%, (trace metal basis), anhydrous, powder, Thermo Scientific Chemicals

168.84 € - 577.88 €

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.6
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10188372
View Documents
Thermo Scientific Acros
378350050
5 g Glass bottle
168.84 €
5g
Please to purchase this item. Need a web account? Register with us today!
10311863
View Documents
Thermo Scientific
378350250
25 g Glass bottle
577.88 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

7718-54-9
129.6
QMMRZOWCJAIUJA-UHFFFAOYSA-L
CHEBI:34887
Cl[Ni]Cl
Cl2Ni
MFCD00011142
24385
dichloronickel

Specifications

1001.0°C
Brown to Yellow
44.6 to 46% (Ni) (Complexometry)
NiCl2
15, 6590
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel
24385
129.6
Trace Metal Basis
7718-54-9
Liquid
Cl2Ni
MFCD00011142
Solubility in water: slightly soluble
Cl[Ni]Cl
129.6
CHEBI:34887
99.99%
Nickel(II) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-743-

Product Identifier
  • Nickel(II) chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only